D-(+)-Trehalose dihydrate ≥99%, white powder cell culture reagent

Supplier: 0000012207

0219475605 0219475610 0219475625 0219475680
IC0219475605EA 56.38 USD
IC0219475605 IC0219475610 IC0219475625 IC0219475680
D-(+)-Trehalose dihydrate ≥99%, white powder cell culture reagent
D(+)-Trehalose dihydrate

Trehalose Dihydrate is an osmolyte, a chemical chaperone. It contains many properties that allow it to stabilize partially unfolded protein molecules and inhibit protein aggregation. It include a low chemical reactivity, high glass transition temperature, existence of numerous polymorphs, nonreducing nature, and high affinity for water molecules. Additionally, Trehalose has been noted to bind to expanded polyglutamines and stabilize partially unfolded polyglutamine-containing proteins.


  • Storage temperature: Room Temperature (15 to 30 °C)


Trehalose, Dihydrate, is an inducer of autophagy, it protect cells against numerous environmental stresses. It is seldom used as a direct replacement for conventional sweeteners. Use as a cryoprotectant in a variety of cell freezing media.

Formula: C₁₂H₂₂O₁₁·2H₂O
Melting Pt: 203 °C (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00071594
CAS Number: 6138-23-4
EINECS: 202-739-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=af741ee934bfe7c82395d31c21e2c19f0738d0f0&vid=59183c854dcfa367e15d46861ef4b9a7028540a9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Physical Appearance White Powder
Solubility Soluble in water (50 mg/mL - clear, colorless solution) or hot alcohol.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR