D-(+)-Galactose, anhydrous ≥98%, white powder, Cell Culture Grade

Supplier: 0000012207

0219467080 0219467090 0219467091
IC0219467080EA 92.25 USD
IC0219467080 IC0219467090 IC0219467091
D-(+)-Galactose, anhydrous ≥98%, white powder, Cell Culture Grade
D(+)-Galactose

  • Grade : Anhydrous, Cell culture reagent
  • Purity : ≥98%
  • Suitability : Suitable for cell culture, insect cell culture
  • Product Type : Biochemicals
  • Biochemical Category : Carbohydrates
  • Chemical Class : Carbohydrates
  • Optical Rotation : +77° ± 5° (c=4, water)
  • Presentation : White powder
  • Format : Powder
  • Detection Method : FTIR
  • Moisture content : ≤1.0%
  • Solubility : Soluble in water (100 mg/mL - clear, colorless solution).

Formula: C₆H₁₂O₆
Melting Pt: 168...170 °C
Storage Temperature: Ambient
MDL Number: MFCD00151230
CAS Number: 59-23-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=91743c4ad898f1896f6821aba6c8f62ff13862c8&vid=91743c4ad898f1896f6821aba6c8f62ff13862c8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
Purity 98%
FTIR Conforms to Standard
Melting Point 160 - 170°C
Moisture 1.0%
Optical Rotation (c=4, water) +77° ± 5°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR