Cetrimonium bromide ∼98% for molecular biology
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
|
Danger
|
Synonyms:
N-Cetyl-N,N,N-trimethylammonium bromide, Cetyltrimethylammonium bromide, Hexadecyltrimethylammonium bromide, CTAB, (1-Hexadecyl)trimethylammonium bromide, N,N,N-Trimethylhexadecan-1-aminium bromide
Cetyltrimethylammonium bromide is a phase transfer catalyst ideal for the precipitation of DNA. Hexadecyltrimethylammonium bromide is a bactericidal, cationic detergent. Its activity is neutralized by soaps and anionic detergents (i.e., SDS). Trimethylammonium bromide compounds form insoluble complexes with SDS. It is active at alkaline pH against both Gram positive and Gram negative organisms.
- Storage temperature: Room Temperature (15 to 30 °C)
Cetyltrimethylammonium bromide ia used as an cationic surfactant suitable for the isolation of high molecular weight DNA in plants and other organisms (by a rapid method) as well as plant DNA for use in PCR analysis. It has been used to precipitate nucleic acids. CTAB provides enhancement of Concanavalin A mediated agglutination. It has been used for the determination of protein molecular weights in electrophoretic systems and for determination of critical micelle concentration of detergents. CTAB has also been used as a titrant for potentiometric titration of perchlorate and as a phase-transfer catalyst in reduction of arenes and heterocyclic compounds.
Formula:
CH₃(CH₃)₁₅N(Br)(CH₃)₃ Melting Pt: 248…251 °C (lit.) Flash Pt: 244 °C (lit.) |
MDL Number:
MFCD00011772 CAS Number: 57-09-0 EINECS: 200-311-3 UN: 3077 ADR: 9,III |
Specification Test Results
Physical Appearance | Powder |
pH | 6.0 - 7.5 (0.1 M H2O, 0 °C)(Lit.) |
Aggregation Number | 170 |
Critical Micelle Concentration | 1 mM (20-25°C) |
Solubility | Freely but slowly soluble in water; Freely soluble in methanol and ethanol; soluble in alcohol. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...