(S)-(+)-2-(6-Methoxy-2-naphthyl)propionic acid

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

Synonyms: Naproxen, (S)-2-(6-Methoxynaphthalen-2-yl)propanoic acid, (S)-(+)-6-Methoxy-alpha-methyl-2-naphthaleneacetic Acid

0219024705
IC0219024705EA 68.14 USD
IC0219024705
(S)-(+)-2-(6-Methoxy-2-naphthyl)propionic acid
(S)-(+)-2-(6-Methoxy-2-naphthyl)propionic acid

Non-selective cyclooxygenase (COX-1 and COX-2) inhibitor. Naproxen has been demonstrated to block Aβ fibril growth and form a complex with copper (II).


Pharmacological agent that reportedly may show activity through ability to inhibit prostaglandin biosynthesis.

Formula: C₁₄H₁₄O₃
MW: 230.26 g/mol
Boiling Pt: 403.9 °C (760 mmHg)) (lit.)
Melting Pt: 152…160 °C
Density: 1,20 g/cm³ (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00010500
CAS Number: 22204-53-1
EINECS: 244-838-7
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Physical Appearance White Powder
Vapor Pressure 3.01E-07 mmHg at 25 °C (lit.)
Optical Rotation (c=2, Ethanol) +59° to +62°
Solubility Soluble in water (3 mg/mL) at 25 °C, PBS pH 7.2, SOL., ethanol, DMSO, DMF (100 mg/mL), chloroform, dichloromethane, ether, ethyl acetate, methanol, and THF. Soluble in acetone (50 mg/mL).

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR