Polyvinylpyrrolidone K30, white powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

Synonyms: Povidone, Polyvinylpyrrolidon, Polyvidone, PVP

02102786.1 02102786.5 0210278605
IC02102786.1EA 126.03 USD
IC02102786.1 IC02102786.5 IC0210278605
Polyvinylpyrrolidone K30, white powder
Polyvinylpyrrolidone

Polyvinylpyrrolidone K 30 is used as thickeners, dispersing agents, detoxicant, complexing agent, lubricants and binders. Polyvinylpyrrolidone is a component of Denhardt′s Solution.


  • Presentation: White Powder
  • Tickeners
  • Dispersing Agents
  • Detoxicant
  • Complexing Agent

Formula: (C₆H₉NO)n
Density: 1.23…1.29 g/mL (lit.)
Storage Temperature: Ambient
MDL Number: MFCD01076626
CAS Number: 9003-39-8

Order Now

Specification Test Results

Identity Test Passes
FTIR Conforms to Standard
pH (5% aq soln) 2.8-4.2
Viscocity (1% aq soln) 27-35cps
Solubility (2.5% aq soln) Clearly Soluble, Free of Floating Particles
Aldehydes 500ppm max
Peroxide 400ppm max
Formic Acid 0.5% max
Hydrazine 1ppm max
Vinylpyrrolidone 10ppm max
2-Pyrrolidone 3.0% max
Lead 10ppm max
Water 5.0% max
Residue on Ignition 0.1% max
Assay (Nitrogen Content) 11.5%-12.8% (Anhydrous Basis)
Total Plate Count 1000cfu/g max
Staph Negative
Salmonella Negative
E.coli Negative
Yeast and Mold 100cfu/g max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR