Malonic acid ≥99%, white crystalline powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Danger

0210223880
IC0210223880EA 98.81 USD
IC0210223880
Malonic acid ≥99%, white crystalline powder
Malonic acid

Malonic acid is used to prepare barbiturates. Its derivative diethylmalonate is used in Knoevenagel condensation reactions as well as reacts with acetone to form Meldrum's acid. It is used to control acidity, preservative additive for foods and as an excipient in pharmaceutical formulation. It acts as a precursor to polyesters and as an active component in alkyd resins. Further, it is used as a building block in chemical synthesis. In addition to this, it is used to prepare starch-based resin.


  • Presentation: White Crystalline Powder
  • Soluble in acetone, benzene, carbon tetrachloride, ether, methanol and water (50 mg/mL-clear, colorless solution).

Formula: C₃H₄O₄
Melting Pt: 132…139 °C
Storage Temperature: Ambient
MDL Number: MFCD00002707
CAS Number: 141-82-2

Order Now

Specification Test Results

Identity Test Passed
Purity ≥99%
FTIR Conforms to Standard


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR