Glycine ≥99.5%

Supplier: IBI Scientific

Synonyms: Aminoacetic acid, Glycocoll, H-Gly-OH

IB70194
IB70194EA 148.96 USD
IB70194
Glycine ≥99.5%
Glycine

Glycine is a neutral amino acid and is a primary component in polyacrylamide buffers.


  • A280 (1M Water): <0.10
  • Heavy metals: <0.002%
  • Loss on Drying: ≤0.2%
  • pH (10% water): 5.5-6.5
  • Protease/ RNAse: none detected
  • Solubility (10% water): Pass


During the process of separation, glycine forms an electrical front which pulls the macromolecules along in a gel run.


Store at room temperature, keep tightly sealed, and protect from moisture.

Formula: NH₂CH₂COOH
Boiling Pt: 181 °C (1013 hPa)
Melting Pt: 83 °C
Density: 1.64 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008131
CAS Number: 56-40-6
EINECS: 200-272-2
Merck Index: 13,04504
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d944007d30d5d4650062014a1b152bd9bfb66ea3&vid=093f4227cbdd1c4c6515ef9c4b85d93799877dcc&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR