EDTA Solution, IBI Scientific
Supplier: IBI Scientific
IB70185
IB70185EA 132.45 USD
IB70185
EDTA Solution, IBI Scientific
Buffers Cloning Buffers
EDTA Solution is commonly used in biological and electrophoresis buffer systems.
Store at room temperature and protect from moisture.
A280-600 < 0.10%