HR2025 Horizontal Gel Electrophoresis System, IBI Scientific
Supplier: IBI Scientific
IB56000
IB56000EA 982.83 USD
IB56000
HR2025 Horizontal Gel Electrophoresis System, IBI Scientific
Electrophoresis Systems Horizontal Gel Electrophoresis Systems
Ideal for inversion and genomic DNA separations
Electrical gravity connector minimizes electrical hazards and eliminates sample disturbance during setup. Gold-plated electrical fittings and 99.9% platinum electrodes resist corrosion and withstand rapid, high voltage polarity switching applications.
Buffer recirculation capability permits high resolution electrophoresis of nucleic acids in low ionic strength buffers. Dedicated casting system eliminates the need for tape and permits gel preparation while minimizing leaks. Well visualization stripes improve visibility to facilitate gel loading, even with a sample quick loader. Vented lid reduces condensation, improving resolution of runs. UVT gel tray with multiple comb slots allows maximum sample capacity using several combs, or maximum running lengths using a single comb. Tray transmits 312nm UV light.
Ordering information: System includes a buffer chamber; safety cover and power cords; casting system and tray; leveling bubble; buffer recirculator port set; two 2.0mm thick, 20-tooth combs; and owner’s manual.