Dithiothreitol (DTT, Cleland's reagent)

Supplier: IBI Scientific
Warning

IB21040
IB21040EA 97.2 USD
IB21040
Dithiothreitol (DTT, Cleland's reagent)
Dithiothreitol (DTT, Cleland's reagent)

DTT is used in enzyme preparations to prevent formation of disulfides from sulfhydryl groups, thereby stabilizing the enzyme.


Caution: Store at -20°C and keep tightly sealed. Warm to room temperature before opening. A DTT solution should be stored in small aliquots at -20°C. It is advisable that DTT solutions should be made fresh prior to use. Do not autoclave DTT or solutions containing DTT.

Formula: C₄H₁₀O₂S₂
MDL Number: MFCD00004877
CAS Number: 3483-12-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5022472c3f3baef75a703791c548fc6504df810f&vid=81529732db015cb66c1f106092309ae5714325b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

-SH content Min. 99.4%
A283 (0.02 M, H2O) Max. 0.05
-S-S-content Max. 0.5%
Melting Point 41 ± 2°C
IR Pass
Moisture Max 0.5%
Solubility (5%, water) Clear and Colorless


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR