Glycerine ≥99.5%, Molecular Biology Grade
Supplier: IBI Scientific
Glycerol is used to prevent freezing during enzyme and bacterial preparations.
Dnase, Rnase, and Protease Free
Caution: Store at room temperature, protect from moisture, and keep tightly sealed.
Formula:
HOCH₂CH(OH)CH₂OH Boiling Pt: 290 °C (1013 hPa) Melting Pt: 18.6 °C Density: 1.26 g/cm³ (20 °C) Flash Pt: 177 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004722 CAS Number: 56-81-5 EINECS: 200-289-5 Merck Index: 13,04497 |
|
Specification Test Results
Purity | >99.5% |
Sulfate | <0.0001% |
Arsenic | <0.0003% |
Chlorine | <0.0003% |
Iron | <0.0005% |
Residue on Ignition | <0.01% |
Density | 1.249-1.262 g/ml |
Specific Gravity | 1.2635 – 1.2615 |
DNase Assay | None Detected |
RNase Assay | None Detected |
Protease Assay | None Detected |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...