Sodium dodecyl sulfate (SDS) ≥99.0%

Supplier: IBI Scientific

IB07062
IB07062EA 169.25 USD
IB07062
Sodium dodecyl sulfate (SDS) ≥99.0%
Sodium dodecyl sulphate (SDS)

SDS is an ionic detergent used to denature proteins in hybridization, nucleic purification, and electrophoresis buffer systems. SDS is also used to dissociate nucleic acid protein complexes in DNA extraction protocols, to disrupt cell membranes and to prepare prehybridization and/or hybridization solutions.


  • % of straight chain C12 SDS: >98
  • Chloride: <0.1%
  • Copper: <0.0005%
  • Heavy metals as Pb: ≤0.0005%
  • Phosphate: ≤0.0005%


Dnase-, Rnase, Protease- Free. Store at room temperature and protect from moisture.

Formula: CH₃(CH₂)₁₁OSO₃Na
Melting Pt: 204…207 °C
Density: 1.1 g/cm³ (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00036175
CAS Number: 151-21-3
EINECS: 205-788-1
UN: 1325
ADR: 4.1,II
Merck Index: 14,08636
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe4cb1741fba41f699a3be006d0b01b1b551898c&vid=c6df9a86fc351b319e68cc79c1c17f733b4fec67&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR