Select-D and NuClean™ Spin Columns, IBI Scientific
Supplier: IBI Scientific
IB06000 IB06005 IB06010 IB06015 IB06020 IB06030 IB06035 IB06040 IB06045 IB06060 IB06080 IB06050 IB06055 IB06065
IB06000PK 301.56 USD
IB06000 IB06005 IB06010 IB06015 IB06020 IB06030 IB06035 IB06040 IB06045 IB06060 IB06080 IB06050 IB06055 IB06065
Select-D and NuClean™ Spin Columns, IBI Scientific
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
Select-D Columns are ideal for desalting and removing unincorporated radiolabeled deoxynucleotide triphosphates from end labels, fill in, nick-translation, or random primed DNA labeling reactions
The complete operation takes a total of ten minutes. Columns come fully hydrated and ready to use. All Select-D columns are RNase and DNase free.