Guanidinium thiocyanate ≥99.0%

Supplier: IBI Scientific

Synonyms: Guanidinium sulfocyanide, Guanidinium sulphocyanide, GTC, Guanidine Thiocyanate, Guanidinium rhodanide

IB05100
IB05100EA 257.53 USD
IB05100
Guanidinium thiocyanate ≥99.0%
Guanidinium thiocyanate

Guanidine Thiocyanate is used in the isolation of RNA.


Dnase, Rnase, and Protease Free. Store at room temperature, tightly sealed, protect from moisture and light.

Formula: CH₅N₃·CHNS
Melting Pt: 118…121 °C
MDL Number: MFCD00013027
CAS Number: 593-84-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR