Guanidinium hydrochloride

Supplier: IBI Scientific

IB05080
IB05080EA 175.92 USD
IB05080
Guanidinium hydrochloride
Guanidinium hydrochloride

Guanidine Hydrochloride is an ionic denaturant that rapidly and effectively denatures most proteins


Guanidine HCl is available as a solid or solution. The solution is composed of 6M Guanidine HCl and 25mM EDTA at pH 7.5.


Dnase, Rnase, and Protease Free. Store at room temperature and protect from moisture.


It is also used for isolation of RNA.

Formula: NH₂C(=NH)NH₂·HCl
MDL Number: MFCD00013026
CAS Number: 50-01-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da5a871cab977e6d9dc666b457732eb1e869a714&vid=b0d4fcb044b6106fce8e40448dbf2ce6e7077ad7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR