X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside)

Supplier: IBI Scientific

IB02265 IB02260 IB02264
97060-740EA 859.96 USD
97060-740 IB02260 IB02264
X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside)
X-Gal (5-Bromo-4-chloro-3-indolyl-ß-D-galactopyranoside)

X-GAL is a histochemical substrate for β-galactosidase, allowing the detection of lac gene expression


Store at -20°C, keep tightly sealed, and warm to room temperature prior to opening.


To make a stock solution of X-GAL, dissolve the X-GAL in dimethylformamide to make a 20mg/ml solution.

Formula: C₁₄H₁₅BrClNO₆
MDL Number: MFCD00005666
CAS Number: 7240-90-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR