Streptomycin sulfate

Supplier: IBI Scientific

IB02180
IB02180EA 49.94 USD
IB02180
Streptomycin sulfate
Streptomycin sulphate

Streptomycin Sulfate is used to bind to S12 protein and the 30S ribosomal subunit


Refrigerated shipping and storage is required for this product.


It inhibits translocation and elicits miscoding bactericidal. Streptomycin is used at the following concentrations: Stringent plasmids (10µg/ml) and Relaxed Plasmids (50µg/ml). Antibiotic inhibition/sensitivity assay, Crystallinity and Chromatographic Purity: PASS.

Formula: C₂₁H₃₉N₇O₁₂·1.5H₂SO₄
MW: 1457.4 g/mol
MDL Number: MFCD00037023
CAS Number: 3810-74-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR