Ampicillin sodium salt for molecular biology

Supplier: IBI Scientific

Synonyms: D(-)-α-Aminobenzylpenicillin Sodium Salt, Ampicillin Na, Ampicillin sodium salt, D-(−)-α-Aminobenzylpenicillin sodium salt Ampicillin sodium salt

IB02040
IB02040EA 106.36 USD
IB02040
Ampicillin sodium salt for molecular biology
Ampicillin sodium salt
Formula: C₁₆H₁₈N₃NaO₄S
MW: 371.39 g/mol
MDL Number: MFCD00064313
CAS Number: 69-52-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR