Boc-Leu-Arg-Arg-AMC · 2 HCl
Supplier: BACHEM AMERICAS INC
4017087.0050
I-1585.0050BAEA 873.19 USD
I-1585.0050BA
Boc-Leu-Arg-Arg-AMC · 2 HCl
Proteins and Peptides
Substrates for measuring the chymotryptic, PGPH, and tryptic acitivity of the 20S or 26S proteasome. See also the thrombin substrate I-1140.
Boc-LLR-AMC is a good substrate for the Kex2 endoprotease (kexin, previously known as proteinase yscF), a membrane-bound, calcium-dependent serine protease from yeast α-cells. Boc-LRR-AMC is also used for determining the trypsin-like activity of the proteasome.