Neuropeptide Y (22-36) Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4015553.0005
H-9305.0005BAEA
1047.05
USD
H-9305.0005BA
Neuropeptide Y (22-36) Trifluoroacetate
Proteins and Peptides
This C-terminal fragment of NPY is a more potent inhibitor of calmodulin-stimulated phosphodiesterase than NPY itself. The inhibitory effect obviously depends on the basic amphiphilic helical nature of the fragment.
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)