Human Relaxin H2 Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4039910.1000
H-6784.0200BAEA 783.89 USD
H-6784.0200BA H-6784.1000BA
Human Relaxin H2 Trifluoroacetate
Proteins and Peptides
Relaxin H2 is the major stored and circulating isoform of the peptide hormone relaxin in humans. This pregnancy hormone has also shown cardioprotective activity. The observed long-term effects of relaxin H2 on connective tissues may be due to alterations in the turnover of collagen and proteoglycans.