Crustacean Erythrophore Concentrating Hormone
Supplier: BACHEM AMERICAS INC
4013731.0005
H-6750.0005BAEA 818.92 USD
H-6750.0005BA
Crustacean Erythrophore Concentrating Hormone
Proteins and Peptides
Crustacean erythrophore concentrating hormone, pELNFSPGWamide, is also called red pigment concentrating hormone (RPCH). This crustacean hormone, which was first isolated from the eyestalk of the pink shrimp Pandalus borealis, has been detected in many decapod species.