Dendroaspis Natriuretic Peptide
Supplier: BACHEM AMERICAS INC
4033517.0500
H-4904.0500BAEA 571.91 USD
H-4904.0500BA
Dendroaspis Natriuretic Peptide
Proteins and Peptides
The 38 amino acid peptide DNP, which has been originally isolated from the venom of the green mamba snake (Dendroaspis angusticeps), shares structural homology with the family of natriuretic peptides. DNP contains a 17 amino acid disulfide ring similar to ANP, BNP, and CNP, which mediate biological actions through particulate guanylyl cyclase receptors and generation of guanosine monophosphate (cGMP).