(D-Trp⁶)-LHRH-Leu-Arg-Pro-Gly amide Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4029116.0001
H-4582.0001BAEA
555.54
USD
H-4582.0001BA
(D-Trp⁶)-LHRH-Leu-Arg-Pro-Gly amide Trifluoroacetate
Proteins and Peptides
LHRH is also called Gonadotropin-Releasing Hormone (GnRH) or Luteinizing Hormone-Releasing Factor (LRF).
H-4075 corresponds to triptorelin acetate, H-6150 to triptorelin pamoate (triptorelin embonate).
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)