(D-Phe⁶,Leu-NHEt¹³,des-Met¹⁴)-Bombesin (6-14) Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4030433.0005
H-3042.0005BAEA 441.71 USD
H-3042.0005BA
(D-Phe⁶,Leu-NHEt¹³,des-Met¹⁴)-Bombesin (6-14) Trifluoroacetate
Proteins and Peptides
DPDMB, bombesin receptor antagonist with IC₅₀ values for cellular secretion and growth approaching the picomolar region.