(N-Me-Tyr¹,N-Me-Arg⁷,D-Leu-NHEt⁸)-Dynorphin A (1-8) Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4030565.0500
H-3018.0500BAEA 473.84 USD
H-3018.0500BA
(N-Me-Tyr¹,N-Me-Arg⁷,D-Leu-NHEt⁸)-Dynorphin A (1-8) Trifluoroacetate
Proteins and Peptides
The N-terminus of dynorphin, YGGFL, corresponds to Leu-enkephalin. So, dynorphin A (1-5) amide is filed as Leu-enkephalin amide (H-2745). For other opioid peptides see, the product families ‘Deltorphins and Dermorphins’, ‘Endorphins, MPF, Neoendorphins’, and ‘Opioid Peptides'.
Highly stable dynorphin-like analgesic peptide.