(R)-(+)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid

Supplier: BACHEM AMERICAS INC

Synonyms: (R)-1,2,3,4-Tetrahydro-3-isoquinolinecarboxylic Acid, H-D-Tic-OH, D-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid

4016023.0001 4016023.0005
F-2585.0001BAEA 114.5 USD
F-2585.0001BA F-2585.0005BA
(R)-(+)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid
(R)-(+)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid

Tic represents a constrained Phe analog.

Formula: C₁₀H₁₁NO₂
Storage Temperature: Refrigerator
MDL Number: MFCD00144038
CAS Number: 103733-65-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR