L-α-Aminoadipic acid

Supplier: BACHEM AMERICAS INC

Synonyms: L-2-Aminoadipic acid, (S)-2-Aminoadipic Acid

4014920.0001 4014920.0005 4014920.0025
F-2560.0001BAEA 117.38 USD
F-2560.0001BA F-2560.0005BA F-2560.0025BA
L-α-Aminoadipic acid
L-α-Aminoadipic acid

Sequence: H-Aad-OH
Synonym(s): L-α-Aminoadipic acid#(S)-2-Aminohexanedioic acid#H-Homoglu-OH


The gliotoxic homolog of glutamic acid is a product of lysine metabolism in mammalian cells. Aad is a precursor of penicillin biosynthesis. A marker of protein oxidation in human skin.

Formula: C₆H₁₁NO₄
MW: 161.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00002636
CAS Number: 1118-90-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR