4-Nitro-DL-phenylalanine

Supplier: BACHEM AMERICAS INC

Synonyms: H-DL-Phe(4-NO₂)-OH, H-p-Nitro-DL-Phe-OH, H-DL-Phe(4-NO2)-OH

4000106.0005
F-1905.0005BAEA 114.5 USD
F-1905.0005BA
4-Nitro-DL-phenylalanine
4-Nitro-DL-phenylalanine

Sequence: H-p-Nitro-DL-Phe-OH


Bachem offers a considerable choice of ring-substituted phenylalanines, protected derivatives of: p-Aminophenylalanine p-Azidophenylalanine p-Benzoylphenylalanine p-Biphenylalanine p-Bromophenylalanine p-t-Butylphenylalanine p-Carboxyphenylalanine p-Chlorophenylalanine p-Cyanophenylalanine 3,4-Dichlorophenylalanine Fluorophenylalanines p-Iodophenylalanine p-Methylphenylalanine p-Nitrophenylalanine p-Phosphonophenylalanine. p-Methoxy- and p-ethoxyphenylalanine derivatives are listed in the Tyr family, whereas β,β-diphenylalanine derivatives can be found in the "β-Substituted Alanines" family.
p-Nitrophenylalanine (4-Nph, Phe(4-NO₂)) is incorporated into peptides as a chromophore. In FRET substrates, it acts as quencher. Bachem also offers resin-bound Fmoc-Nph (Fmoc-p-nitro-Phe-Wang resin, D-1135).

Formula: C₉H₁₀N₂O₄
MW: 210.189 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00007384
CAS Number: 2922-40-9
EINECS: 220-868-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR