D-Cycloserine
Supplier: BACHEM AMERICAS INC
Sequence: D-Cycloserine
Synonym(s): (R)-4-Aminoisoxazalidin-3-one
Originally, D-cycloserine has been used as an antibiotic in the treatment of tuberculosis. More recently, it has found application in treating anxiety disorders (see the review by S.G.Hofmann et al.). Acting as a partial agonist of the NMDA receptor, the compound reduces neuropathic pain.
Formula:
C₃H₆N₂O₂ Storage Temperature: Freezer |
MDL Number:
MFCD00005353 CAS Number: 68-41-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...