Vacuum Gauge, Dial, Series CG16K, Edwards
Supplier: Edwards
D35610000
EVD356-10-000EA 1542.37 USD
EVD356-10-000
Vacuum Gauge, Dial, Series CG16K, Edwards
Pumps Vacuum Gauges
This gauge is barometrically independent and ruggedly constructed
Easy-to-read linear scale. Suitable for panel mounting or direct pipeline mounting via NW16 flange. Accuracy is ±2% of full scale deflection