Anti-Fos (34E4) Mouse Monoclonal Antibody [clone: 34E4]

Supplier: MilliporeSigma
ST1029-1SET
EMST1029-1SETEA 720.01 USD
EMST1029-1SET
Anti-Fos (34E4) Mouse Monoclonal Antibody [clone: 34E4]
Antibodies
Primary Mouse Anti-Fos (pSer 374 ) (34E4) Reacts with Human

Type: Primary
Antigen: Fos (pSer 374 ) (34E4)
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR