Live/Dead Cell Staining Kit, MilliporeSigma
Supplier: MilliporeSigma
QIA76-100TEST
EMQIA76100TESTEA 874.12 USD
EMQIA76100TEST
Live/Dead Cell Staining Kit, MilliporeSigma
Assays Cellular Assays
The Live/Dead Cell Staining Kit utilizes two fluorescent dyes, Cyto-dye to stain live cells and propidium iodide to stain dead cells.
  • Utilizes two fluorescent dyes
  • Band-pass filter which detects FITC and rhodamine
Stained live and dead cells can be visualized by fluorescence microscopy using a band-pass filter which detects FITC and rhodamine. Viable cells will stain only with the Cyto-dye, fluorescing green, whereas the dead cells will stain with both Cyto-dye (green) and propidium iodide (red), resulting in a yellow fluorescence.