Anti-SYK Rabbit Polyclonal Antibody

Supplier: MilliporeSigma
PK1010-50UL
EMPK1010-50ULEA 401.9 USD
EMPK1010-50UL
Anti-SYK Rabbit Polyclonal Antibody
Antibodies
Primary Rabbit Anti-Syk (pTyr 525/526 ) Reacts with Human, Mouse

Type: Primary
Antigen: SYK
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR