Anti-MDM2 (Ab-1) Mouse Monoclonal Antibody [clone: IF2]

Supplier: MilliporeSigma
OP46-100UG
EMOP46-100UGEA 526.61 USD
EMOP46-100UG
Anti-MDM2 (Ab-1) Mouse Monoclonal Antibody [clone: IF2]
Antibodies
Purified mouse monoclonal antibody (see application references). Recognizes the ~90 kDa (apparent MW) MDM2 protein. Also recognizes isoforms at ~57 and ~74/76 kDa.

Type: Primary
Antigen: MDM2 (Ab-1)
Clonality: Monoclonal
Clone: IF2
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR