Anti-IKKα Mouse Monoclonal Antibody [clone: 14A231]

Supplier: MilliporeSigma
OP133-100UG
EMOP133-100UGEA 649.94 USD
EMOP133-100UG
Anti-IKKα Mouse Monoclonal Antibody [clone: 14A231]
Antibodies
This Anti-IKKα Mouse mAb (14A231) is validated for use in Immunoblotting for the detection of IKKα.

Type: Primary
Antigen: IKKα
Clonality: Monoclonal
Clone: 14A231
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Monkey;Human;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR