Anti-5-mC Mouse Monoclonal Antibody

Supplier: MilliporeSigma
NA81-50UG
EMNA81-50UGEA 471.78 USD
EMNA81-50UG
Anti-5-mC Mouse Monoclonal Antibody
Antibodies
Primary Mouse Anti-5-Methylcytosine (162 33 D3) Reacts with Pan

Type: Primary
Antigen: 5mC
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: pan
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR