Xylene (mixture of isomers) ≥98.5% (xylene isomers plus ethylbenzene, GC), GR ACS, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Dimethylbenzene

XX0055-5 XX0055P-4 XX0055-3 XX0055-20 XX0055P-4 XX0055-5 XX0055-6
CAXX0055-5CS 4178.18 USD
CAXX0055-5 CAXX0055P-4 EM-XX0055-3 EM-XX0055-20 EM-XX0055P-4 EMD-XX0055-5 EM-XX0055-6
Xylene (mixture of isomers) ≥98.5% (xylene isomers plus ethylbenzene, GC), GR ACS, Supelco®
Xylene (mixture of isomers)

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs

Formula: C₆H₄(CH₃)₂
MW: 106.17 g/mol
Boiling Pt: 137…140 °C (lit.)
Melting Pt: –34 °C
Density: 0.86…0.88 g/cm3 (25 °C)
Flash Pt: 25 °C
Storage Temperature: Ambient
MDL Number: MFCD00077264
CAS Number: 1330-20-7
EINECS: 215-535-7
UN: 1307
ADR: 3,III
Merck Index: 13,10136
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2deefc7b7b67c10e64003e243ff3216fad27cf7d&vid=9fa01ddafd4814d5f34266ae7933463a29d92d19&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (xylene isomers plus ethylbenzene, GC) 98.5% min
Color (APHA) 10 max
Ethylbenzene 25% max
Form Clear liquid
Residue after evaporation 0.002% max
Substances darkened by sulfuric acid To pass test
Sulfur compounds (as S) 0.003% max
Water (H2O) 0.05% max

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR