Trifluoroacetic acid 0.1% (v/v) in water mobile phase for HPLC, Supelco®

Supplier: MilliporeSigma

Synonyms: TFA

TX1280P-1
EMD-TX1280P-1CS 1613 USD
EMD-TX1280P-1
Trifluoroacetic acid 0.1% (v/v) in water mobile phase for HPLC, Supelco®
Trifluoroacetic acid
Formula: C₂HF₃O₂
MW: 114.02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00004169
CAS Number: 76-05-1
EINECS: 200-929-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9501f3ac85d66e8d19d1cd1c3fe5409b974e9d8a&vid=be6aa627129b49ccda76006e7003b46d79b933cc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (trifluoroacetic acid) 0.095-0.105% (v/v)
Color (APHA) 10 max
Filtered through 0.2 µm filter To pass test
Form Clear liquid
Gradient at 210 nm 0.01AU max
Gradient at 220 nm 0.001AU max
Gradient at 280 nm 0.001AU max
Residue after evaporation 5ppm max
UV Abs. at 205 nm 1.00AU max
UV Abs. at 210 nm 0.70AU max
UV Abs. at 230 nm 0.30AU max
UV Abs. at 250 nm 0.05AU max
UV Abs. at 280 nm 0.010AU max
UV Abs. at 300 nm 0.010AU max
UV Abs. at 350 nm 0.010AU max

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR