Sulfuric acid 95.0-98.0%, GR ACS ACS, Supelco®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

SX1244-5 SX1244-6 SX1244-75 SX1244-23 SX1244-75 SX1244PC-5 SX1244-6 SX1244-5 SX1244-23
CASX1244-5CS 1445.52 USD
CASX1244-5 CASX1244-6 CASX1244-75 EM-SX1244-23 EM-SX1244-75 EM-SX1244PC-5 EM-SX1244-6 EMD-SX1244-5 TXSX124423WRE
Sulfuric acid 95.0-98.0%, GR ACS ACS, Supelco®
Sulfuric acid

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs.

Formula: H₂SO₄
MW: 98.08
Boiling Pt: 330 (lit.)
Melting Pt: 10.38
Density: 1.840 g/ml (25°C) (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 1830
ADR: 8 II

Order Now

Specification Test Results

Aluminum (Al) 0.2ppm max
Ammonium (NH4) 2ppm max
Appearance To pass test
Arsenic (As) 0.01ppm max
Assay 95.0-98.0%
Calcium (Ca) 0.3ppm max
Chloride (Cl) 0.2ppm max
Chromium (Cr) 0.2ppm max
Color (APHA) 10 max
Copper (Cu) 0.1ppm max
Gold (Au) 0.3ppm max
Heavy metals (as Pb) 1ppm max
Iron (Fe) 0.2ppm max
Lead (Pb) 0.3ppm max
Magnesium (Mg) 0.3ppm max
Manganese (Mn) 0.2ppm max
Mercury (Hg) 5ppb max
Nickel (Ni) 0.1ppm max
Nitrate (NO3) 0.5ppm max
Potassium (K) 0.3ppm max
Residue after ignition 5ppm max
Sodium (N/A) 0.3ppm max
Substances reducing permangaN/Ate (as SO2) 2ppm max
Tin (Sn) 0.2ppm max
Titanium (Ti) 0.3ppm max
Zinc (Zn) 0.2ppm max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR