Orthophosphoric acid ≥85% (w/w), GR ACS, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Phosphoric acid

M-Clarity™ quality level = MQ300

PX0995-3 PX0995-4000 PX0995-6 PX0995-78 PX0995P-1 PX0995-4000 PX0995-6 PX0995P-5 PX0995-3
CAPX0995-3CS 3963.3 USD
CAPX0995-3 CAPX0995-4000 CAPX0995-6 EM-PX0995-78 EMD-PX0995P-1 EM-PX0995-4000 EM-PX0995-6 EM-PX0995P-5 EMD-PX0995-3
Orthophosphoric acid ≥85% (w/w), GR ACS, Supelco®
Orthophosphoric acid

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs

Formula: H₃PO₄
MW: 98 g/mol
Melting Pt: 28 °C
Density: 1.71…1.87 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011340
CAS Number: 7664-38-2
EINECS: 231-633-2
UN: 1805
ADR: 8,III

Order Now

Specification Test Results

Antimony (Sb) 0.002% max
Arsenic (As) 1ppm max
Assay 85.0% min
Calcium (Ca) 0.002% max
Chloride (Cl) 3ppm max
Color (APHA) 10 max
Form Clear liquid
Heavy metals (as Pb) 0.001% max
Insoluble matter 0.001% max
Iron (Fe) 0.003% max
Magnesium (Mg) 0.002% max
Manganese (Mn) 0.5ppm max
Nitrate (NO3) 5ppm max
Potassium (K) 0.005% max
Reducing substances To pass test
Sodium (N/A) 0.025% max
Sulfate (SO4) 0.003% max
Volatile acids (as acetic acid) 0.001% max

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR