Methanol ≥99.8% (by GC), GR ACS, meets analytical specification for testing USP/NF monographs, Supelco®
Supplier: MilliporeSigma
|
Danger
|
MX0485-5
MX0485-3
MX0485-7
MX0485-8
MX0485-9
MX0485-8
MX0485-5
MX0485-6
MX0485-9
MX0485-3
MX0485-20
MX0485-7
MX0485P-4
CAMX0485-5EA
1441.92
USD
CAMX0485-5
CAMX0485-3
CAMX0485-7
CAMX0485-8
CAMX0485-9
EM-MX0485-8
EM-MX0485-5
EM-MX0485-6
EM-MX0485-9
EM-MX0485-3
EM-MX0485-20
EMD-MX0485-7
EMD-MX0485P-4
EMMX0485-31
Methanol ≥99.8% (by GC), GR ACS, meets analytical specification for testing USP/NF monographs, Supelco®
Methanol
Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs.
Formula:
CH₃OH Boiling Pt: 64.7 °C (lit.) Melting Pt: –98 °C (lit.) Density: 0.791 g/ml (25°C) (lit.) Flash Pt: 11 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00004595 CAS Number: 67-56-1 EINECS: 200-659-6 UN: 1230 ADR: 3,6.1,II REACH: 01-2119433307-44 Merck Index: 13,05984 |
|
Specification Test Results
Appearance | Clear |
Assay (GC) | 99.8% min |
Carbonyl (acetone;formaldehyde;acetaldehyde) | 0.001% max |
Color (APHA) | 10 max |
Evaporation residue | 0.001% max |
Solubility in water | To pass test |
Substances darkened by sulfuric acid | To pass test |
Substances reducing permanganate | To pass test |
Titrable acid | 0.0003meq/g max |
Titrable base | 0.0002meq/g max |
Water (H2O) | 0.1% max |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...