Ethanol denatured 94.0-96.0% (v/v), GR ACS ACS for histology, for biochemistry (denatured with 5% isopropanol and 5% methanol), Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Ethyl alcohol denatured

AX0441-5 AX0441-7 AX0441-6 AX0441-3 AX0441-20 AX0441-20
EM-AX0441-5EA 1164 USD
EM-AX0441-5 EM-AX0441-7 EM-AX0441-6 EMD-AX0441-3 EM-AX0441-36 TXLAX044120WNG EM-AX0441-20
Ethanol denatured 94.0-96.0% (v/v), GR ACS ACS for histology, for biochemistry (denatured with 5% isopropanol and 5% methanol), Supelco®
Ethanol denatured

Used as the final stage in dehydration for processing tissue especially in electron microscopy.Prior to shipment of unit sizes of Reagent Alcohol over 4L, a letterhead application approved by the Bureau of Alcohol, Tobacco & Firearms (ATF) must be on file


  • Total alcohol: ~100%
  • Ethyl alcohol: 89-92%
  • Denaturant: 4-6% methyl alcohol and 4-6% 2-propanol


Clear, colorless, flammable liquid, hydrophilic and miscible with water. Used as the final stage in dehydration for processing tissue especially in electron microscopy.


Ordering information: The purchase of Reagent Alcohol in containers larger than 4 liters requires a Letterhead Permit approval by the United States Treasury Tax and Trade Bureau (TTB).

Formula: H₃CCH₂OH
MW: 46.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003568
CAS Number: 64-17-5
EINECS: 200-578-6
UN: 1987
ADR: 3 II

Order Now

Specification Test Results

Assay (ethanol + methanol) 94.0-96.0% (v/v)
Assay (ethanol) 89.0-92.0% (v/v)
Assay (isopropyl alcohol) 4.0-6.0% (v/v)
Assay (methanol) 3.5-5.5% (v/v)
Color (APHA) 10 max
Form Clear liquid
Infrared spectrum Conforms to standard
Residue after evaporation 0.001% max
Water (H2O) 0.075% max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR