Anti-ICAM-1 (W-CAM-1) Mouse Monoclonal Antibody

Supplier: MilliporeSigma
CP53-100UG
EMCP53-100UGEA 483.06 USD
EMCP53-100UG
Anti-ICAM-1 (W-CAM-1) Mouse Monoclonal Antibody
Antibodies
Primary Mouse Anti-ICAM-1 (Ab-2) (W-CAM-1) Reacts with Human

Type: Primary
Antigen: ICAM-1 (Ab-2) (W-CAM-1)
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR