Pressure Reducing Valve, MilliporeSigma
Supplier: MilliporeSigma
9.67100.9100
EM9.67100.9100EA 1434 USD
EM9.67100.9100
Pressure Reducing Valve, MilliporeSigma
Valves Pressure-Relief Valves
The pressure reducer with included over-pressure relief is one of the safest combined solutions for reducing inert gas house lines to a working pressure of 0.2 bar.
The integrated overpressure relieve avoids pressures higher than 0.5 bar inside the barrel according the pressure equipment directive. The pressure reducer is equipped with a ball valve and with two tube connectors 8×6 mm.
Packaging: Packed in a plastic bag.