Dosing Unit for Liquid products in the Extran® Range
Supplier: MilliporeSigma
9.57571.1020
EM9.57571.1020EA 15 USD
EM9.57571.1020
Dosing Unit for Liquid products in the Extran® Range
Cleaning Agents Detergents and Surface Cleansers
To support accurate dosage and safe handling in manual cleaning, MilliporeSigma supplies 1 liter Extran® bottles with a special dosing aid. The bottle and attachment are perfectly matched to facilitate dosage. Since neither too little nor too much detergent is used, you benefit from reliable cleaning as well as economical use. The reusable Extran® dosing aid can also be ordered separately if required.
  • High convenience: The system is easy to handle and greatly facilitates dosage
  • Safety: Accurate dosage of detergents prevents excess chemicals in the workplace
  • Economy: The dosing aid provides precise and accurate dosing and avoids unnecessary expenditure of cleaning agents