Water for synthesis, deionized, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.48333.9910
EM8.48333.1000CS 200 USD
EM8.48333.1000 EM8.48333.9010 EM8.48333.9200 EM8.48333.9910
Water for synthesis, deionized, Sigma-Aldrich®
Water
Formula: H₂O
Boiling Pt: 100 °C (lit.)
Melting Pt: 0 °C (lit.)
Density: 1.000 g/ml at 3.98 °C (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Order Now

Specification Test Results

Boiling point 99 - 101 °C
Density (d 20 °C/ 4 °C) 0.9979 - 0.9985
Conductivity (20 °C) ≤ 4.3 µS/cm
- -


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR