(4-Carboxybutyl)triphenylphosphonium bromide for pharma synthesis, non-GMP, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
M-Clarity™ quality level = MQ400
8.45134.0025
8.45134.0050
EM8.45134.0025CS
6929
USD
EM8.45134.0025
EM8.45134.0050
EM8.45134.1000
(4-Carboxybutyl)triphenylphosphonium bromide for pharma synthesis, non-GMP, Sigma-Aldrich®
(4-Carboxybutyl)triphenylphosphonium bromide
Formula:
C₂₃H₂₄BrO₂P Melting Pt: 205…208 °C |
MDL Number:
MFCD00011906 CAS Number: 17814-85-6 EINECS: 241-782-5 UN: 1325 ADR: 4.1,III |
Specification Test Results
Solubility | (20 °C) soluble |
Melting Point | 204 - 208 °C |
Molar Mass | 443.31 g/mol |
Bulk Density | 360 - 400 kg/m3 |
Flash Point | 195 °C |
Hazard Class | 6.1 |
Hazard Class Description | Toxic materials |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...