(4-Carboxybutyl)triphenylphosphonium bromide for pharma synthesis, non-GMP, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ400

8.45134.0025 8.45134.0050
EM8.45134.0025CS 6929 USD
EM8.45134.0025 EM8.45134.0050 EM8.45134.1000
(4-Carboxybutyl)triphenylphosphonium bromide for pharma synthesis, non-GMP, Sigma-Aldrich®
(4-Carboxybutyl)triphenylphosphonium bromide
Formula: C₂₃H₂₄BrO₂P
Melting Pt: 205…208 °C
MDL Number: MFCD00011906
CAS Number: 17814-85-6
EINECS: 241-782-5
UN: 1325
ADR: 4.1,III

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 204 - 208 °C
Molar Mass 443.31 g/mol
Bulk Density 360 - 400 kg/m3
Flash Point 195 °C
Hazard Class 6.1
Hazard Class Description Toxic materials


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR