Benzethonium chloride, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
Hyamine® 1622, N-Benzyl-N,N-dimethyl-2-(2-(4-(2,4,4-trimethylpentan-2-yl)phenoxy)ethoxy)ethanaminium chloride
M-Clarity™ quality level = MQ200
8.43983.0250
EM8.43983.0250CS
2795
USD
EM8.43983.0250
Benzethonium chloride, Sigma-Aldrich®
Benzethonium chloride
Benzethonium chloride for synthesis
Formula:
C₂₇H₄₂ClNO₂ MW: 448.09 g/mol |
MDL Number:
MFCD00011742 CAS Number: 121-54-0 UN: 2923 ADR: 8 (6.1),III |
Specification Test Results
Hill Formula | C₂₇H₄₂ClNO₂ |
Molar Mass | 448.08 g/mol |
Ignition temperature | 380 °C |
Melting Point | 160 - 166 °C |
Vapor pressure |
|
Bulk density | 440 kg/m³ |
Solubility | (20 °C) soluble |
LD 50 oral | LD50 Rat 295 mg/kg |
Signal Word | Danger |
Storage class | 6.1C Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects |
WGK | WGK 2 water endangering |
R Phrase | R 22-34-50/53 |
S Phrase | S 26-36/37/39-45-61 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...